missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic acid hydrazide, 90%, Thermo Scientific Chemicals

$191.79 - $666.81

Chemical Identifiers

CAS 624-84-0
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
MDL Number MFCD00007589
InChI Key XZBIXDPGRMLSTC-UHFFFAOYSA-N
Synonym formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
PubChem CID 12229
IUPAC Name formohydrazide
SMILES C(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376060250
SDS View Documents
Thermo Scientific Chemicals
376060250
25 g Glass bottle
Each for $191.79
 
AC376061000
SDS View Documents
Thermo Scientific Chemicals
376061000
100 g Glass bottle
Each for $666.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

624-84-0
60.06
XZBIXDPGRMLSTC-UHFFFAOYSA-N
12229
C(=O)NN
CH4N2O
MFCD00007589
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
formohydrazide

Specifications

624-84-0
White
Authentic
Glass bottle
HCONHNH2
25 g
Solubility in water: soluble in water
C(=O)NN
60.06
60.06
Shiny Crystalline Flakes
51°C to 58°C
112°C
90%
CH4N2O
MFCD00007589
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
XZBIXDPGRMLSTC-UHFFFAOYSA-N
formohydrazide
12229
90%
Formic acid hydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-867-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only