missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formic acid, for mass spectrometry, Honeywell Fluka™
for mass spectrometry, ∼98%
$263.70 - $263.70
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
MDL Number | MFCD00003297 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Specifications
64-18-6 | |
HCOOH | |
UN1779 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
98%, 98% (T) | |
250 mL | |
49°C (120.2°F) | |
1.6 (vs air) | |
Formic acid |
CH2O2 | |
MFCD00003297 | |
1209246 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.03g/mol | |
1.22g/mL at 25°C | |
0.57 | |
1004°F | |
44.8 mmHg (20°C) |
Safety and Handling
P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
RTECSNumber : LQ4900000