missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formohydrazide 97.0+%, TCI America™
Supplier: TCI America F0667100G
Specifications
Formohydrazide | |
59°C | |
90°C | |
MFCD00007589 | |
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine | |
C(=O)NN | |
60.056 | |
60.06 | |
Crystalline Powder |
624-84-0 | |
White | |
CH4N2O | |
100 g | |
XZBIXDPGRMLSTC-UHFFFAOYSA-N | |
formohydrazide | |
12229 | |
≥97.0% (HPLC) |
Chemical Identifiers
624-84-0 | |
60.056 | |
XZBIXDPGRMLSTC-UHFFFAOYSA-N | |
12229 | |
C(=O)NN |
CH4N2O | |
MFCD00007589 | |
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine | |
formohydrazide |
Safety and Handling
RTECSNumber : LQ8615000
TSCA : Yes
Recommended Storage : Refrigerator