missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formononetin, 98%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 342480010

Catalog No. AC342480010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Formononetin
485-72-3
97.5% min. (HPLC)
C16H12O4
1g
formononetin, biochanin b, formononetol, 7-hydroxy-4'-methoxyisoflavone, 7-hydroxy-3-4-methoxyphenyl-4h-chromen-4-one, 4'-o-methyldaidzein, 7-hydroxy-3-4-methoxyphenyl chromen-4-one, unii-295dqc67bj, 7-hydroxy-3-4-methoxyphenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-methoxyphenyl
COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
268.27
CHEBI:18088
98%
99%
Authentic
Glass bottle
MFCD00016948
15, 4271
HKQYGTCOTHHOMP-UHFFFAOYSA-N
7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
5280378
268.27

Chemical Identifiers

485-72-3
268.27
HKQYGTCOTHHOMP-UHFFFAOYSA-N
5280378
7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
C16H12O4
MFCD00016948
formononetin, biochanin b, formononetol, 7-hydroxy-4'-methoxyisoflavone, 7-hydroxy-3-4-methoxyphenyl-4h-chromen-4-one, 4'-o-methyldaidzein, 7-hydroxy-3-4-methoxyphenyl chromen-4-one, unii-295dqc67bj, 7-hydroxy-3-4-methoxyphenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-methoxyphenyl
CHEBI:18088
COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-623-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only