missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Formylmethyl)triphenylphosphonium Chloride 98.0+%, TCI America™
$61.51 - $175.22
Chemical Identifiers
| CAS | 62942-43-2 |
|---|---|
| Molecular Formula | C20H18ClOP |
| Molecular Weight (g/mol) | 340.79 |
| MDL Number | MFCD00012003 |
| InChI Key | RVEJRPJGKXTQIF-UHFFFAOYSA-M |
| Synonym | formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride |
| PubChem CID | 2733863 |
| IUPAC Name | (2-oxoethyl)triphenylphosphanium chloride |
| SMILES | [Cl-].O=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 62942-43-2 | |
| 340.79 | |
| RVEJRPJGKXTQIF-UHFFFAOYSA-M | |
| 2733863 | |
| [Cl-].O=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H18ClOP | |
| MFCD00012003 | |
| formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride | |
| (2-oxoethyl)triphenylphosphanium chloride |
Specifications
| 62942-43-2 | |
| MFCD00012003 | |
| formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride | |
| [Cl-].O=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 340.79 | |
| 340.79 | |
| Crystalline Powder |
| C20H18ClOP | |
| 5 g | |
| RVEJRPJGKXTQIF-UHFFFAOYSA-M | |
| (2-oxoethyl)triphenylphosphanium chloride | |
| 2733863 | |
| ≥98.0% (T) | |
| (Formylmethyl)triphenylphosphonium Chloride |
Safety and Handling
TSCA : No