Foss™ Thimbles for Soxtec™ 2055 Manual Fat Extraction System
33mm Cellulose Thimbles
Specifications
Material Cellulose
For Use With (Application) Used for fat extraction
For Use With (Equipment) For soxtec 2055
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
TC1522-0045
View Documents
Foss™
15220045
Extraction Thimble, Single Thickness N/A
Description
Description
Specifications
Specifications
Cellulose
Used for fat extraction
For soxtec 2055
SDS
missing translation for 'documents'
missing translation for 'documents'