Bulwark™ Women's FR/CP Lab Coats
Lightweight and breathable lab coat provides dual hazard protection with flame resistance and chemical protection.
$337.56 - $354.43
Specifications
Product Type Flame Resistant Lab Coat
Gender Women's
Material Nomex IIIA
Color Royal Blue
Closure Type Snaps
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19800068
View Documents
Bulwark™
KNR3RBSM
Small
Each for $354.43
19800067
View Documents
Bulwark™
KNR3RBMD
Medium
Each for $354.43
19800066
View Documents
Bulwark™
KNR3RBLG
Large
Each for $337.56
Description
Description
  • Nomex™ IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
  • Nomex™ knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
  • Black lay-down collar
  • Concealed snap front closure
  • Splash-resistant side pass-through for easy inner pocket access
  • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
  • Available in a variety of sizes
  • Includes a waistband in the back for a more tailored fit
Certifications
NFPA 2112 UL-Certified
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire. UL certified.
Specifications
Specifications
Flame Resistant Lab Coat
Nomex IIIA
Snaps
Reusable
Flame
3
354CH45RB
128 g
Women's
Royal Blue
Knit
Non-sterile
Regular
Westex ShieldCXP Chemical Splash Protection
4.5 oz.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'