missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Freezer Storage Racks

Organize and optimize the freezers in the lab.

Supplier:  Fisherbrand™ 398267

 View more versions of this product

Catalog No. 03-395-466


Explore available promotions

Description

Description

Organize and optimize the freezers in the lab

Make it easy to track samples by improving organization in your freezers. These racks enable you to store more samples, eliminate dead space, and minimize frost buildup.

  • Fit all standard types and sizes of upright and chest freezers, regardless of manufacturer
  • Store a variety of sample containers, including boxes, plates, and tubes
  • High-quality, corrosion resistant stainless steel
  • For use in all standard upright and chest freezers

Specifications

Specifications

Freezer Storage Rack: Side Access Rack
Boxes (2 in.)
20
Upright Freezers
68.1 cm
23.9 cm
5.3 in.
2 in.
Not Autoclavable
Stainless Steel
20
20
26.8 in.
9.4 in.
13.5 cm
Videos
SDS
Documents

Documents

Promotions

Promotions