missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vileda Professional™ Buckets and Downpress Wringer Combination
Vileda Professional™'s bucket and wringer combos are designed to meet the demanding requirements of the most sanitary environments.
$220.66 - $268.66
Specifications
Product Type | Bucket and Wringer |
---|---|
Material | Plastic |
Color | Yellow |
For Use With (Application) | Designed to meet the demanding requirements of the most sanitary environments |
For Use With (Equipment) | Fits all mop sizes |
Description
- Quiet, smooth operation, easy to work with
- More efficient wringing, avoids water spalshing on the floor
- Accommodates all wet mop sizes with convenient opening
- Durable, stain resistant one piece construction is easy to clean and sanitize and does not provide a breeding ground for bacteria
- Downward pressure wringer forces water into bucket more effectively
Specifications
Bucket and Wringer | |
Yellow | |
Fits all mop sizes |
Plastic | |
Designed to meet the demanding requirements of the most sanitary environments | |
Gears and pressure plates with Fiber reinforced nylon, single bail carry handle, durable ball bearing casters, downward pressure wringer |