Honeywell™ North™ RU6500 Series Full-Facepiece Air-Purifying Respirators
Features a polycarbonate lens with a wide field of vision for maximized peripheral vision plus a downward view.
Supplier:  Honeywell™ RU65001L
 View more versions of this product
Catalog No. 19-085-554
Explore available promotions
Requires : Compatible with North™ N-Series cartridges, filters and accessories.
Description
Description
  • Silicone seal provides secure facepiece seal, creating a barrier against contaminants
  • Silicone nose cup is soft and comfortable for all day wear
  • Wide viewing area provides distortion free visibility and optical clarity
  • Polycarbonate lens meet ANSI Z.87 for high impact
  • Facepiece with 5-point strap includes center strap to help position and hold the mask for a more secure fit; offers quick, easy adjustments
  • Facepiece with optional industrial nylon mesh head net (19-085-555, 556, 557) provides another choice for user comfort preference; can be used under hard hat
  • Threaded cartridge connector secures equipment to avoid accidental disconnections
  • Can be adapted to side- or front-mounted PAPR or CF-SAR; compatible with PA700 Series PAPR; PR500 Series Mask-Mounted PAPR; CF2000 Series SAR; and CF4000 Series Mask-Mounted SAR
Specifications
Specifications
ANSI Z87.1
Silicone
Large
North™ N-Series line of cartridges, filters, and cartridge/filter combinations
Polycarbonate
5-point
Black
Full Facepiece Respirator
Silicone
Head Strap
RU65001L