ILC Dover Full Skirt Hood for Sentinel XT Blower
Full skirt hood has an APF of 1,000 and can be used with the Sentinel XL and XT Blower Systems.
$1412.36 - $3505.32
Specifications
Assigned Protection Factor (APF) | 1000 |
---|---|
Product Type | Full Skirt Hood |
For Use With (Application) | Healthcare, Pharmaceutical |
For Use With (Equipment) | Sentinel XT Blower |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
19-631-077
|
ILC Dover
S311310 |
10/Pk. |
Pack of 10 Each for $1,412.36
|
|
|||||
19631078
|
ILC Dover
S311325 |
25/Pk. |
Each of 25 for $3,505.32
|
|
|||||
Description
- Ideal for both healthcare and pharmaceutical industries
Specifications
1000 | |
Full Skirt Hood | |
Healthcare, Pharmaceutical | |
Sentinel XT Blower |