Learn More
Fullerene C60, 99.9%, Thermo Scientific Chemicals
$149.13 - $359.97
Chemical Identifiers
CAS | 99685-96-8 |
---|---|
Molecular Formula | C60 |
Molecular Weight (g/mol) | 720.66 |
MDL Number | MFCD00151408 |
InChI Key | XMWRBQBLMFGWIX-UHFFFAOYSA-N |
Synonym | fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih |
PubChem CID | 123591 |
ChEBI | CHEBI:33128 |
IUPAC Name | (C\{60}-I\{h})[5,6]fullerene |
SMILES | C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC295012500
|
Thermo Scientific Chemicals
295012500 |
250 mg | Glass bottle |
Each for $149.13
|
|
||||
AC295010010
|
Thermo Scientific Chemicals
295010010 |
1 g | Glass bottle |
Each for $359.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99685-96-8 | |
720.66 | |
XMWRBQBLMFGWIX-UHFFFAOYSA-N | |
123591 | |
(C\{60}-I\{h})[5,6]fullerene |
C60 | |
MFCD00151408 | |
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih | |
CHEBI:33128 | |
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41 |
Specifications
99685-96-8 | |
Authentic | |
Glass bottle | |
MFCD00151408 | |
15, 1472 | |
Solubility in water: insoluble. Other solubilities: >5mg/mL in toluene | |
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41 | |
720.66 | |
CHEBI:33128 | |
99.90% | |
Fullerene C60, 99.9% |
Black to Brown | |
99.85% min. (HPLC) | |
C60 | |
250 mg | |
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih | |
XMWRBQBLMFGWIX-UHFFFAOYSA-N | |
(C\{60}-I\{h})[5,6]fullerene | |
123591 | |
720.65 | |
Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only