missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fullerene powder, sublimed, 99.9+% C{60}, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 042008MF

 View more versions of this product

Catalog No. AA42008MF

Explore available promotions

Description

Description

The optical absorption properties of C60 match solar spectrum in a way that suggests that C60-based films could be useful for photovoltaic applications. Because of its high electronic affinity it is one of the most common electron acceptor used in donor/acceptor based solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fullerene powder
99685-96-8
1.7 g/mL
C60
MFCD00151408
UN1325
Insoluble in water. Soluble in organic solvents.
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
720.66
CHEBI:33128
Powder
Sublimed
Black
500°C to 600°C (sublimation)
C60
50 mg
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
XMWRBQBLMFGWIX-UHFFFAOYSA-N
(C\{60}-I\{h})[5,6]fullerene
123591
≥99.9%

Chemical Identifiers

99685-96-8
720.66
XMWRBQBLMFGWIX-UHFFFAOYSA-N
123591
(C\{60}-I\{h})[5,6]fullerene
C60
MFCD00151408
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
CHEBI:33128
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : LS9252500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only