Learn More
Fumaric acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1097636
Description
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.
Notes
Incompatible with oxidizing agents, amines and strong bases.
Specifications
Fumaric acid | |
∼299°C (sublimation) | |
2.1 | |
Odorless | |
MFCD00002700 | |
605763 | |
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(E)-but-2-enedioic acid | |
444972 | |
116.07 |
110-17-8 | |
1.625 | |
273°C (523°F) | |
C4H4O4 | |
500 g | |
14,4287 | |
Soluble in water,ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform,olive oil,carbon tetrachloride,xylene,molten camphor,liquid ammonia and benzene. | |
OC(=O)\C=C\C(O)=O | |
116.07 | |
CHEBI:18012 | |
99% |
Chemical Identifiers
110-17-8 | |
116.07 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
444972 | |
(E)-but-2-enedioic acid |
C4H4O4 | |
MFCD00002700 | |
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid | |
CHEBI:18012 | |
OC(=O)\C=C\C(O)=O |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319-H500
EINECSNumber : 203-743-0
RTECSNumber : LS9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only