Learn More
Fumaric acid monoethyl ester, 95%
CAS: 2459-05-4 | C6H8O4 | 144.13 g/mol
$471.65 - $471.65
Chemical Identifiers
| CAS | 5-4-2459 |
|---|---|
| Molecular Formula | C6H8O4 |
| Molecular Weight (g/mol) | 144.13 |
| MDL Number | MFCD00002699 |
| InChI Key | XLYMOEINVGRTEX-ONEGZZNKSA-N |
| Synonym | Monoethylefumarate |
| IUPAC Name | (2E)-4-ethoxy-4-oxobut-2-enoic acid |
| SMILES | CCOC(=O)\C=C\C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC156201000
|
Thermo Scientific Chemicals
156201000 |
100 g | Glass bottle |
Each for $471.65
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5-4-2459 | |
| 144.13 | |
| XLYMOEINVGRTEX-ONEGZZNKSA-N | |
| (2E)-4-ethoxy-4-oxobut-2-enoic acid |
| C6H8O4 | |
| MFCD00002699 | |
| Monoethylefumarate | |
| CCOC(=O)\C=C\C(O)=O |
Specifications
| 5-4-2459 | |
| Beige to White | |
| Authentic | |
| Glass bottle | |
| HO2CCH=CHCO2C2H5 | |
| 100 g | |
| Monoethylefumarate | |
| CCOC(=O)\C=C\C(O)=O | |
| 144.13 | |
| 95% | |
| Fumaric acid monoethyl ester |
| 64°C to 69°C | |
| 147°C (16.0 mmHg) | |
| 95% | |
| C6H8O4 | |
| MFCD00002699 | |
| 02, 741 | |
| XLYMOEINVGRTEX-ONEGZZNKSA-N | |
| (2E)-4-ethoxy-4-oxobut-2-enoic acid | |
| 144.13 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 219-544-7
RUO – Research Use Only