missing translation for 'onlineSavingsMsg'
Learn More

FURA 2-AM

CAS: 108964-32-5 | C44H47N3O24 | 1001.857 g/mol

Supplier:  Thermo Scientific Chemicals J62728MCR

Catalog No. AAJ62728MCR


Only null left
Explore available promotions

Description

Description

A calcium indicator; derivative of Fura-2 used to load the chelator into cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A calcium indicator; derivative of Fura-2 used to load the chelator into cells
Specifications

Specifications

FURA 2-AM
C44H47N3O24
8183748
VPSRLGDRGCKUTK-UHFFFAOYSA-N
acetyloxymethyl 2-[6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
3364574
Light sensitive
1 mg
108964-32-5
MFCD00036976
Fura-2 pentakis(acetoxymethyl) ester
CC1=CC(=C(C=C1)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC2=C(C=C3C(=C2)C=C(O3)C4=NC=C(O4)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C
1001.857
1001.87
Yellow
Powder

Chemical Identifiers

108964-32-5
1001.857
VPSRLGDRGCKUTK-UHFFFAOYSA-N
3364574
CC1=CC(=C(C=C1)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC2=C(C=C3C(=C2)C=C(O3)C4=NC=C(O4)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C
C44H47N3O24
MFCD00036976
Fura-2 pentakis(acetoxymethyl) ester
acetyloxymethyl 2-[6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only