Learn More
Furan, 99+%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 181110010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| Furan | |
| 0.9360g/mL | |
| −35°C | |
| 0.05% max. (Furfural) (GC) | |
| C4H4O | |
| MFCD00003222 | |
| 2389 | |
| 07,161; 09,227; 10,187; 13,173 | |
| 15, 4325 | |
| Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,ether and aceton,hydrolysis in water | |
| O1C=CC=C1 | |
| 68.08 | |
| 8029 | |
| 68.06 |
| 110-00-9,128-37-0 | |
| 31.3°C | |
| Authentic | |
| Glass bottle | |
| 1.4197 to 1.4217 | |
| 1L | |
| 17, 27 | |
| 0.936 | |
| divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane | |
| YLQBMQCUIZJEEH-UHFFFAOYSA-N | |
| furan | |
| 0.025 to 0.04% BHT | |
| CHEBI:35559 | |
| 99+% |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.
May cause cancer.
Harmful to aquatic life with long
GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the
GHS Signal Word: Danger
EINECSNumber : 203-727-3
RUO â Research Use Only