missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Furfural | |
-37°C | |
>3.0 | |
C5H4O2 | |
500 g | |
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural | |
O=CC1=CC=CO1 | |
96.09 | |
CHEBI:34768 | |
≥98.0% (GC) |
98-01-1 | |
Yellow | |
160°C | |
MFCD00003229 | |
1199 | |
HYBBIBNJHNGZAN-UHFFFAOYSA-N | |
furan-2-carbaldehyde | |
7362 | |
96.09 | |
Liquid |
Chemical Identifiers
98-01-1 | |
96.09 | |
HYBBIBNJHNGZAN-UHFFFAOYSA-N | |
7362 | |
furan-2-carbaldehyde |
C5H4O2 | |
MFCD00003229 | |
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural | |
CHEBI:34768 | |
O=CC1=CC=CO1 |
Safety and Handling
EINECSNumber : (5)-0040
RTECSNumber : LT7000000
TSCA : Yes