missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl acetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1528514

 View more versions of this product

Catalog No. AAA1528514

Explore available promotions

Description

Description

Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Furfuryl acetate
1.118
65°C (149°F)
1.463
25 g
furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
CKOYRRWBOKMNRG-UHFFFAOYSA-N
furan-2-ylmethyl acetate
12170
97%
623-17-6
175°C to 177°C
C7H8O3
MFCD00003251
116128
Not miscible in water.
CC(=O)OCC1=CC=CO1
140.138
140.14

Chemical Identifiers

623-17-6
140.138
CKOYRRWBOKMNRG-UHFFFAOYSA-N
12170
CC(=O)OCC1=CC=CO1
C7H8O3
MFCD00003251
furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
furan-2-ylmethyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 210-775-9

RTECSNumber : LU9120000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only