missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

$71.24 - $199.60

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119792500
SDS View Documents
Thermo Scientific Chemicals
119792500
250 mL Glass bottle
Each for $71.24
 
AC119790010
SDS View Documents
Thermo Scientific Chemicals
119790010
1 L Glass bottle
Each for $165.43
 
AC119790025
SDS View Documents
Thermo Scientific Chemicals
119790025
2.5 L Glass bottle
Each for $199.60
Case of 4 Each for $0.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-00-0
98.1
XPFVYQJUAUNWIW-UHFFFAOYSA-N
7361
furan-2-ylmethanol
C5H6O2
MFCD00003252
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
CHEBI:207496
C1=COC(=C1)CO

Specifications

98-00-0
Yellow
170°C
Authentic
Glass bottle
1.4850 to 1.488
250 mL
01,408
15, 4334
Solubility in water: miscible but unstable
C1=COC(=C1)CO
98.1
5 mPa.s (25°C)
98.1
Liquid
-29°C
1.1300g/mL
65°C
97.5% min. (GC)
C5H6O2
MFCD00003252
17, V,3, 338
1.13
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
XPFVYQJUAUNWIW-UHFFFAOYSA-N
furan-2-ylmethanol
7361
CHEBI:207496
98%
Furfuryl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Imm

GHS Signal Word: Danger

EINECSNumber : 202-626-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only