Learn More
Furfuryl glycidyl ether, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 5380-87-0 |
|---|---|
| Molecular Formula | C8H10O3 |
| Molecular Weight (g/mol) | 154.17 |
| MDL Number | MFCD00013348 |
| InChI Key | RUGWIVARLJMKDM-UHFFFAOYNA-N |
| Synonym | furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl |
| PubChem CID | 94306 |
| IUPAC Name | 2-(oxiran-2-ylmethoxymethyl)furan |
| SMILES | C(OCC1=CC=CO1)C1CO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC348490050
|
Thermo Scientific Chemicals
348490050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC348490250
|
Thermo Scientific Chemicals
348490250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5380-87-0 | |
| 154.17 | |
| RUGWIVARLJMKDM-UHFFFAOYNA-N | |
| 94306 | |
| C(OCC1=CC=CO1)C1CO1 |
| C8H10O3 | |
| MFCD00013348 | |
| furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl | |
| 2-(oxiran-2-ylmethoxymethyl)furan |
Specifications
| 5380-87-0 | |
| 103.0°C to 104.0°C (11.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4800 to 1.4820 | |
| 5g | |
| furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl | |
| C(OCC1=CC=CO1)C1CO1 | |
| 154.17 | |
| 154.17 | |
| Furfuryl glycidyl ether |
| 1.1220g/mL | |
| 102°C | |
| 96% min. (GC) | |
| C8H10O3 | |
| MFCD00013348 | |
| 1.122 | |
| RUGWIVARLJMKDM-UHFFFAOYNA-N | |
| 2-(oxiran-2-ylmethoxymethyl)furan | |
| 94306 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May form explosive peroxides.
GHS P Statement:
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 226-372-6
RTECSNumber : LU1423000
RUO â Research Use Only