missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl mercaptan, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2238014

 View more versions of this product

Catalog No. AAB2238014


Explore available promotions

Description

Description

Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Furfuryl mercaptan
1.132
45°C (113°F)
C5H6OS
MFCD00003254
UN1228
Air Sensitive
Insoluble in water.
C1=COC(=C1)CS
114.162
114.17
98-02-2
155°C
Strong
1.5315
25 g
383594
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
ZFFTZDQKIXPDAF-UHFFFAOYSA-N
furan-2-ylmethanethiol
7363
≥98%

Chemical Identifiers

98-02-2
114.162
ZFFTZDQKIXPDAF-UHFFFAOYSA-N
7363
C1=COC(=C1)CS
C5H6OS
MFCD00003254
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
furan-2-ylmethanethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.

EINECSNumber : 202-628-2

RTECSNumber : LU2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only