missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H34400MD

 View more versions of this product

Catalog No. AAH34400MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Furo[3,2-b]pyridine-2-carboxylic acid
>300°C
MFCD11588128
furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
OC(=O)C1=CC2=NC=CC=C2O1
163.13
163.13
112372-14-2
C8H5NO3
250 mg
VHRCHODSRLMXFM-UHFFFAOYSA-N
furo[3,2-b]pyridine-2-carboxylic acid
13803071
97%

Chemical Identifiers

112372-14-2
163.13
VHRCHODSRLMXFM-UHFFFAOYSA-N
13803071
OC(=O)C1=CC2=NC=CC=C2O1
C8H5NO3
MFCD11588128
furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
furo[3,2-b]pyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only