missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
G418 Sulfate Solution, MP Biomedicals
50mg/ml active antibiotic sterile-aqueous solution. Purity: >>98%
$269.75 - $875.35
Specifications
Type | G418 Sulfate |
---|---|
Quantity | 20 mL |
Chemical Name or Material | G418 Sulfate Solution |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN1672546
|
MP Biomedicals Inc
091672546 |
20 mL |
Each for $269.75
|
|
|||||
ICN1672548
|
MP Biomedicals Inc
091672548 |
50 mL |
Each for $875.35
|
|
|||||
Specifications
G418 Sulfate | |
20 mL | |
G418 Sulfate Solution |