Blackline Safety™ G7x Satellite-Connected Safety Monitor with Standard Cartridge (No Gas Detection), North America
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
Supplier:  Blackline Safety™ G7XZNA
Catalog No. 170000142
Explore available promotions
Description
Description
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Specifications
G7 Standard Cartridge
Product Suggestions
Product Suggestions