missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin 98.0+%, TCI America™

$134.21 - $408.22

Chemical Identifiers

CAS 60142-96-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00865286
InChI Key UGJMXCAKCUNAIE-UHFFFAOYSA-N
Synonym gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
PubChem CID 3446
ChEBI CHEBI:42797
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G03185G
View Documents
TCI America
G03185G
5 g
Each for $134.21
Only null left
 
G031825G
View Documents
TCI America
G031825G
25 g
Each for $408.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60142-96-3
171.24
UGJMXCAKCUNAIE-UHFFFAOYSA-N
3446
2-[1-(aminomethyl)cyclohexyl]acetic acid
C9H17NO2
MFCD00865286
gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
CHEBI:42797
C1CCC(CC1)(CC(=O)O)CN

Specifications

166°C
White
MFCD00865286
UGJMXCAKCUNAIE-UHFFFAOYSA-N
2-[1-(aminomethyl)cyclohexyl]acetic acid
171.24
CHEBI:42797
≥98.0% (GC,T)
Gabapentin
60142-96-3
C9H17NO2
gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
C1CCC(CC1)(CC(=O)O)CN
5 g
3446
171.24
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GU6496000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'