missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

$296.89 - $942.07

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12397FF
View Documents
Thermo Scientific Chemicals
012397FF
25 x 25 mm
Each for $296.89
 
AA12397FI
View Documents
Thermo Scientific Chemicals
012397FI
50 x 50 mm
Each for $942.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
23982
gadolinium
Gd
MFCD00011022
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
CHEBI:33375
[Gd]

Specifications

Silver
Gd
UN3178
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
[Gd]
157.25
CHEBI:33375
Air and moisture sensitive
Foil
(REO)
7440-54-2
MFCD00011022
25 x 25 mm
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
99.9%
≈0.12g/25x25mm
Gadolinium foil, 0.025mm (0.001 in.) thick
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-162-2

RTECSNumber : LW3850000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only