missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 12395FF

 View more versions of this product

Catalog No. AA12395FF


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gadolinium foil
7440-54-2
Gd
UN3178
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
[Gd]
157.25
CHEBI:33375
≈1.2g/25x25mm
0.25mm (0.01 in.) thick
(REO)
MFCD00011022
25 x 25 mm
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
99.9%
Foil

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
23982
gadolinium
Gd
MFCD00011022
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
CHEBI:33375
[Gd]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-162-2

RTECSNumber : LW3850000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only