missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 316761000

 View more versions of this product

Catalog No. AC316761000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Specifications

Gadolinium(III) nitrate hexahydrate
White
Crystalline Powder and/or Crystals
99.90%
Gd(NO3)3·6H2O
gadolinium iii nitrate hexahydrate, gadolinium nitrate hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, nitric acid, gadolinium 3+ salt, hexahydrate, gadoliniumnitratehexahydrate, gd.3no3.6h2o, gadolinium 3+ trinitrate hexahydrate, gadolinium nitrate, hexahydrate,, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium iii nitrate hexahydrate trace metals basis
XWFVFZQEDMDSET-UHFFFAOYSA-N
gadolinium(3+);trinitrate;hexahydrate
209258
99.9%
Glass bottle
91°C
19598-90-4
100 g
GdN3O9·6H2O
MFCD00149728
Solubility in water: soluble
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]
451.4
451.4
Trace Metal Basis

Chemical Identifiers

19598-90-4
451.4
XWFVFZQEDMDSET-UHFFFAOYSA-N
209258
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]
GdN3O9·6H2O
MFCD00149728
gadolinium iii nitrate hexahydrate, gadolinium nitrate hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, nitric acid, gadolinium 3+ salt, hexahydrate, gadoliniumnitratehexahydrate, gd.3no3.6h2o, gadolinium 3+ trinitrate hexahydrate, gadolinium nitrate, hexahydrate,, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium iii nitrate hexahydrate trace metals basis
gadolinium(3+);trinitrate;hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May intensify fire; oxidizer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only