Learn More
Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade
CAS: 14017-52-8
$64.44 - $550.78
Chemical Identifiers
| CAS | 14017-52-8 |
|---|---|
| MDL Number | MFCD00150239 |
| Synonym | gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA40571NA
|
Thermo Scientific Chemicals
040571NA |
2 g |
Each for $64.44
|
|
|||||
|
AA40571NC
|
Thermo Scientific Chemicals
040571NC |
10 g |
Each for $154.11
|
|
|||||
|
AA40571NE
|
Thermo Scientific Chemicals
040571NE |
50 g |
Each for $550.78
|
|
|||||
Description
[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14017-52-8 | |
| MFCD00150239 | |
| gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o |
Specifications
| 14017-52-8 | |
| 2 g | |
| UN3211 | |
| Insoluble in water. | |
| Reagent | |
| Odorless |
| Liquid | |
| MFCD00150239 | |
| gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o | |
| 563.69 (455.60 Anhydrous) | |
| 50% w/w aq. soln. | |
| Gadolinium(III) perchlorate hexahydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber : 237-837-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only