missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040289DH

 View more versions of this product

Catalog No. AA40289DH


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gadolinium rod
7440-54-2
Gd
25 mm
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
99.9%
≈0.25g/mm
6.35mm (0.25 in.) dia.
(metals basis excluding Ta)
MFCD00011022
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
[Gd]
157.25
CHEBI:33375
Mylar under Argon
Cast Rod

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
23982
gadolinium
Gd
MFCD00011022
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
CHEBI:33375
[Gd]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-162-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only