missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gadolinium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040289HF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Gadolinium rod | |
7440-54-2 | |
Gd | |
10 mm | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
gadolinium | |
23982 | |
99.9% | |
≈0.25g/mm |
6.35mm (0.25 in.) dia. | |
(metals basis excluding Ta) | |
MFCD00011022 | |
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds | |
[Gd] | |
157.25 | |
CHEBI:33375 | |
Mylar under Argon | |
Cast Rod |
Chemical Identifiers
7440-54-2 | |
157.25 | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
23982 | |
gadolinium |
Gd | |
MFCD00011022 | |
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds | |
CHEBI:33375 | |
[Gd] |
Safety and Handling
EINECSNumber : 231-162-2
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only