missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium silicide, 99.9% (metals basis), Thermo Scientific™

$155.36 - $155.36

Chemical Identifiers

CAS 12134-75-7
Molecular Formula GdH8Si2+3
Molecular Weight (g/mol) 221.484
MDL Number MFCD00144382
InChI Key AOLGUBMBPNHJFD-UHFFFAOYSA-N
PubChem CID 71434213
IUPAC Name gadolinium(3+);silane
SMILES [SiH4].[SiH4].[Gd+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3635604
View Documents
Thermo Scientific Chemicals
3635604
2 g
Each for $155.36
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12134-75-7
221.484
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
GdH8Si2+3
MFCD00144382
71434213
[SiH4].[SiH4].[Gd+3]

Specifications

12134-75-7
2 g
MFCD00144382
[SiH4].[SiH4].[Gd+3]
221.484
213.42
Odorless
(metals basis)
Pieces
GdH8Si2+3
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
71434213
99.9%
Gadolinium silicide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-216-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'