missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP 92.0+%, TCI America™

$485.86 - $1597.78

Chemical Identifiers

CAS 150412-81-0
Molecular Formula C61H64O12
Molecular Weight (g/mol) 989.171
InChI Key GEVBSODSBYNKQA-IMBADGMBSA-N
PubChem CID 11320628
IUPAC Name (2R,3S,4S,5R,6S)-6-[(2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-yl]oxy-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OC4C(C(C(C(O4)COCC5=CC=CC=C5)O)OCC6=CC=CC=C6)OCC7=CC=CC=C7)OCC8=CC=CC=C8)OCC9=CC=CC=C9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M17261G
View Documents
TCI America
M17261G
1 g
Each for $485.86
 
M17265G
View Documents
TCI America
M17265G
5 g
Each for $1,597.78
 
Description

Description

Specifications

Chemical Identifiers

150412-81-0
989.171
11320628
COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OC4C(C(C(C(O4)COCC5=CC=CC=C5)O)OCC6=CC=CC=C6)OCC7=CC=CC=C7)OCC8=CC=CC=C8)OCC9=CC=CC=C9
C61H64O12
GEVBSODSBYNKQA-IMBADGMBSA-N
(2R,3S,4S,5R,6S)-6-[(2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-yl]oxy-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-ol

Specifications

150412-81-0
C61H64O12
LacMP 4′C-OH, per OBn, 4-Methoxyphenyl 4-O-(2,3,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside
COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OC4C(C(C(C(O4)COCC5=CC=CC=C5)O)OCC6=CC=CC=C6)OCC7=CC=CC=C7)OCC8=CC=CC=C8)OCC9=CC=CC=C9
989.171
989.17
Crystalline Powder
White
1 g
GEVBSODSBYNKQA-IMBADGMBSA-N
(2R,3S,4S,5R,6S)-6-[(2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-yl]oxy-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-ol
11320628
≥92.0% (HPLC)
Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'