Learn More
Galanthamine hydrobromide, Thermo Scientific Chemicals
Centrally active and long-acting acetylcholinesterase inhibitor
Supplier: Thermo Scientific Chemicals J62482MF
Description
Centrally active and long-acting acetylcholinesterase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Galanthamine hydrobromide | |
White | |
MFCD00067672 | |
UN2811 | |
galanthamine hydrobromide, galantamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide | |
QORVDGQLPPAFRS-XPSHAMGMSA-N | |
368.27 | |
368.27 |
1953-04-4 | |
C17H22BrNO3 | |
50 mg | |
14,4340 | |
Soluble in water or DMSO | |
CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br | |
121587 | |
Powder |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 217-780-5
RTECSNumber : DF8075000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only