missing translation for 'onlineSavingsMsg'
Learn More

Galena, naturally occurring mineral, approximately 1-2in

CAS: 1314-87-0 | PbS | 239.30 g/mol

$49.59 - $159.59

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name sulfanylidenelead
SMILES [S--].[Pb++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42546KF
View Documents
Thermo Scientific Chemicals
042546KF
1 Ea.
Each for $49.59
Only null left
 
AA42546KJ
View Documents
Thermo Scientific Chemicals
042546KJ
5 Pc.
Each for $159.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-87-0
239.30
FILXVLQXZYXBSJ-UHFFFAOYSA-N
14819
[S--].[Pb++]
PbS
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
sulfanylidenelead

Specifications

1314-87-0
1 Ea.
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
FILXVLQXZYXBSJ-UHFFFAOYSA-N
sulfanylidenelead
14819
Bag
Crystal Clusters
PbS
UN3077
Soluble in strong HNO3,in excess of hot HCl
[S--].[Pb++]
239.30
239.25
Galena
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 215-246-6

RTECSNumber : OG4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only