missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
H-B Instrument™ Galileo Thermometer
Handcrafted and individually calibrated.
Supplier: H-B Instrument™ B620000200EMD
Description
- Multi-colored spheres become more or less buoyant as the temperature of the liquid they are in changes
- Each sphere differs in weight by only 0.002g
- Filled with nonhazardous liquid
- Standard models use optical crystal and are accurate to ±1°F
- Economy models use laboratory-grade glass and are accurate to ±2°F
Specifications
24 in. | |
24 in. | |
Glass | |
100°F | |
60°F | |
60°F to 100°F | |
1°C |
610 mm | |
610 mm | |
Non Hazardous Liquid | |
32°C | |
16°C | |
16°C to 32°C | |
2°C |