Learn More
Gallic Acid 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 410860010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gallic acid | |
253.0°C | |
Yellow to Beige | |
C7H6O5 | |
gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech | |
LNTHITQWFMADLM-UHFFFAOYSA-N | |
3,4,5-trihydroxybenzoic acid | |
370 | |
170.12 | |
1.6940g/mL | |
Authentic | |
Powder |
98% | |
149-91-7 | |
98% | |
MFCD00002510 | |
Solubility in water: 12g/L cold water. Other solubilities: easily soluble in warmed water, very soluble in ethanol: 160g/L, acetone: 200g/l, soluble in glycerol: 100g/L, slightly soluble in, ether: 10g/L, insoluble in benzene, chloroform, petrol ether | |
OC(=O)C1=CC(O)=C(O)C(O)=C1 | |
170.12 | |
CHEBI:30778 | |
98% | |
1 kg | |
Plastic bottle | |
1.694 |
Chemical Identifiers
149-91-7 | |
170.12 | |
LNTHITQWFMADLM-UHFFFAOYSA-N | |
370 | |
3,4,5-trihydroxybenzoic acid |
C7H6O5 | |
MFCD00002510 | |
gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech | |
CHEBI:30778 | |
OC(=O)C1=CC(O)=C(O)C(O)=C1 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 205-749-9
RTECSNumber : LW7525000
TSCA : TSCA
RUO – Research Use Only