missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallic acid monohydrate, Thermo Scientific Chemicals

$182.15 - $269.92

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
IUPAC Name 3,4,5-trihydroxybenzoic acid;hydrate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC220761000
SDS View Documents
Thermo Scientific Chemicals
220761000
100 g Plastic bottle
Each for $182.15
 
AC220765000
SDS View Documents
Thermo Scientific Chemicals
220765000
500 g Plastic bottle
Each for $269.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5995-86-8
169.11
LNTHITQWFMADLM-UHFFFAOYSA-M
24721416
OC1=CC(=CC(O)=C1O)C([O-])=O
C7H5O5
MFCD00149098
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
3,4,5-trihydroxybenzoic acid;hydrate

Specifications

251.0°C
White to Beige
(HO)3C6H2CO2H·H2O
10, 470
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
LNTHITQWFMADLM-UHFFFAOYSA-M
3,4,5-trihydroxybenzoic acid;hydrate
24721416
≥99% (on dry substance)
Authentic
Plastic bottle
Gallic acid monohydrate, c.p.
5995-86-8
C7H5O5
MFCD00149098
15, 4375
Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, glycerol and acetone, practically insoluble in benzene, chloroform and, petroleum ether
OC1=CC(=CC(O)=C1O)C([O-])=O
169.11
188.12
100 g
10% max. (120°C)
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only