missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Gallic Acid, Saturated, Ricca Chemical
Chemical Identifiers
CAS |
5995-86-8 |
Molecular Formula |
C7H5O5 |
Molecular Weight (g/mol) |
169.11 |
MDL Number |
MFCD00149098 |
InChI Key |
LNTHITQWFMADLM-UHFFFAOYSA-M |
Synonym |
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 |
PubChem CID |
24721416 |
IUPAC Name |
3,4,5-trihydroxybenzoate |
SMILES |
OC1=CC(=CC(O)=C1O)C([O-])=O |
View More Specs
Products
2
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
R3224999500
View Documents
|
Ricca Chemical Company
R3224999500A
|
500 mL |
Natural Poly Bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
|
R32249991A
View Documents
|
Ricca Chemical Company
R32249991A
|
1 L |
Natural Poly Bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
|
Description
Specifications
Chemical Identifiers
|
5995-86-8 |
|
169.11 |
|
LNTHITQWFMADLM-UHFFFAOYSA-M |
|
24721416 |
|
OC1=CC(=CC(O)=C1O)C([O-])=O |
|
C7H5O5 |
|
MFCD00149098 |
|
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 |
|
3,4,5-trihydroxybenzoate |
Specifications
|
5995-86-8,7732-18-5 |
|
C7H5O5 |
|
gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 |
|
OC1=CC(=CC(O)=C1O)C([O-])=O |
|
169.11 |
|
500 mL |
|
Natural Poly Bottle |
|
Saturated |
|
MFCD00149098 |
|
LNTHITQWFMADLM-UHFFFAOYSA-M |
|
3,4,5-trihydroxybenzoate |
|
24721416 |
|
Odorless |
|
Gallic Acid Solution |
Videos
SDS
missing translation for 'documents'