Learn More
Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
$151.40 - $1082.92
Chemical Identifiers
CAS | 13450-91-4 |
---|---|
Molecular Formula | GaI3 |
Molecular Weight (g/mol) | 450.436 |
MDL Number | MFCD00016107 |
InChI Key | DWRNSCDYNYYYHT-UHFFFAOYSA-K |
Synonym | gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis |
PubChem CID | 83478 |
IUPAC Name | triiodogallane |
SMILES | [Ga](I)(I)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA4714803
|
Thermo Scientific Chemicals
04714803 |
1 g |
Each for $151.40
|
|
||||||
AA4714806
|
Thermo Scientific Chemicals
04714806 |
5 g |
Each for $315.93
|
|
||||||
AA4714814
|
Thermo Scientific Chemicals
04714814 |
25 g |
Each for $1,082.92
|
|
||||||
Description
It is useful for the preparation of compounds of gallium(I) and gallium(II) and is reported as useful in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
13450-91-4 | |
450.436 | |
DWRNSCDYNYYYHT-UHFFFAOYSA-K | |
83478 | |
[Ga](I)(I)I |
GaI3 | |
MFCD00016107 | |
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis | |
triiodogallane |
Specifications
345°C (sublimation) | |
13450-91-4 | |
Beads | |
GaI3 | |
UN3260 | |
Decomposes in water. | |
[Ga](I)(I)I | |
450.436 | |
450.43 | |
Ampule | |
4.15 g/mL | |
(metals basis) |
212°C | |
Yellow to Green | |
1 g | |
MFCD00016107 | |
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis | |
DWRNSCDYNYYYHT-UHFFFAOYSA-K | |
triiodogallane | |
83478 | |
99.999% | |
Light and moisture sensitive | |
Gallium(III) iodide, Ultra dry |
Safety and Handling
GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H334-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 236-611-6
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only