Learn More
Gallium(III) nitrate solution, Ga 9-10% w/w
CAS: 13494-90-1 | GaN3O9 | 255.735 g/mol
$245.38 - $963.86
Chemical Identifiers
| CAS | 13494-90-1 |
|---|---|
| Molecular Formula | GaN3O9 |
| Molecular Weight (g/mol) | 255.735 |
| MDL Number | MFCD00011016 |
| InChI Key | CHPZKNULDCNCBW-UHFFFAOYSA-N |
| Synonym | gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt |
| PubChem CID | 61635 |
| IUPAC Name | gallium;trinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3] |
Description
Gallium(III) nitrate solution, is used as the material for producing gallium salts and for scientific research and chemical reagent. It is also used as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13494-90-1 | |
| 255.735 | |
| CHPZKNULDCNCBW-UHFFFAOYSA-N | |
| 61635 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3] |
| GaN3O9 | |
| MFCD00011016 | |
| gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt | |
| gallium;trinitrate |
Specifications
| 13494-90-1 | |
| 50 g | |
| MFCD00011016 | |
| gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt | |
| CHPZKNULDCNCBW-UHFFFAOYSA-N | |
| gallium;trinitrate | |
| 61635 | |
| Ga 9 to 10% w/w |
| Liquid | |
| GaN3O9 | |
| UN3218 | |
| It is soluble in water. | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3] | |
| 255.735 | |
| 255.73 | |
| Gallium (III) nitrate solution |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H271-H290-H314-H331
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
EINECSNumber : 236-815-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only