missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallocyanine, Thermo Scientific Chemicals

$67.77 - $126.47

Chemical Identifiers

CAS 1562-85-2
Molecular Formula C15H13ClN2O5
Molecular Weight (g/mol) 336.72
MDL Number MFCD00011926
InChI Key AQSOTOUQTVJNMY-UHFFFAOYSA-N
Synonym gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
PubChem CID 73801
IUPAC Name 7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
SMILES CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154430100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
154430100
10 g Glass bottle
Each for $67.77
 
AC154430250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
154430250
25 g Glass bottle
Each for $126.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1562-85-2
336.72
AQSOTOUQTVJNMY-UHFFFAOYSA-N
73801
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]
C15H13ClN2O5
MFCD00011926
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride

Specifications

1562-85-2
Authentic
C15H13ClN2O5
10 g
15, 4385
AQSOTOUQTVJNMY-UHFFFAOYSA-N
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
73801
Fine Crystalline Powder
Black to Green
Glass bottle
MFCD00011926
27, 438
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]
336.72
336.72
Gallocyanine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

DOTInformation : DOT Class 6.1

EINECSNumber : 216-345-7

RTECSNumber : SP7692000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only