missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Gas Cylinder Barricade Rack
Justrite™ Gas Cylinder Barricade Rack secures cylinders up to 12-in (305-mm) in diameter
$1072.61 - $5684.03
Specifications
Material | Steel |
---|---|
Color | Black |
For Use With (Equipment) | Gas cylinder |
Height (Metric) | 762 mm |
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
19-085-776
|
Justrite™
35080 |
1 Cylinder: 1 Wide by 1 Deep |
Each for $1,072.61
|
|
|||||
19-085-777
|
Justrite™
35084 |
2 Cylinders: 1 Wide by 2 Deep |
Each for $1,170.56
|
|
|||||
19-085-778
|
Justrite™
35090 |
3 Cylinders: 1 Wide by 3 Deep |
Each for $1,488.79
|
|
|||||
19-085-779
|
Justrite™
35096 |
4 Cylinders: 1 Wide by 4 Deep |
Each for $1,493.84
|
|
|||||
19-085-780
|
Justrite™
35100 |
2 Cylinders: 2 Wide by 1 Deep |
Each for $1,483.10
|
|
|||||
19-085-781
|
Justrite™
35106 |
4 Cylinders: 2 Wide by 2 Deep |
Each for $1,785.78
|
|
|||||
19-085-782
|
Justrite™
35112 |
6 Cylinders: 2 Wide by 3 Deep |
Each for $1,918.36
|
|
|||||
19-085-783
|
Justrite™
35118 |
8 Cylinders: 2 Wide by 4 Deep |
Each for $2,273.61
|
|
|||||
19-085-784
|
Justrite™
35122 |
3 Cylinders: 3 Wide by 1 Deep |
Each for $1,874.71
|
|
|||||
19-085-785
|
Justrite™
35128 |
6 Cylinders: 3 Wide by 2 Deep |
Each for $2,359.58
|
|
|||||
19-085-787
|
Justrite™
35134 |
9 Cylinders: 3 Wide by 3 Deep |
Each for $2,540.06
|
|
|||||
19-085-789
|
Justrite™
35140 |
12 Cylinders: 3 Wide by 4 Deep |
Each for $3,143.99
|
|
|||||
19-085-790
|
Justrite™
35144 |
4 Cylinders: 4 Wide by 1 Deep |
Each for $2,340.39
|
|
|||||
19-085-791
|
Justrite™
35150 |
8 Cylinders: 4 Wide by 2 Deep |
Each for $2,553.82
|
|
|||||
19-085-792
|
Justrite™
35156 |
12 Cylinders: 4 Wide by 3 Deep |
Each for $3,480.76
|
|
|||||
19-085-793
|
Justrite™
35162 |
16 Cylinders: 4 Wide by 4 Deep |
Each for $4,440.65
|
|
|||||
19-085-794
|
Justrite™
35166 |
5 Cylinders: 5 Wide by 1 Deep |
Each for $2,575.58
|
|
|||||
19-085-795
|
Justrite™
35172 |
10 Cylinders: 5 Wide by 2 Deep |
Each for $3,461.85
|
|
|||||
19-085-796
|
Justrite™
35178 |
15 Cylinders: 5 Wide by 3 Deep |
Each for $4,350.95
|
|
|||||
19-085-797
|
Justrite™
35182 |
20 Cylinders: 5 Wide by 4 Deep |
Each for $5,684.03
|
|
|||||
Description
- Meets USGS high-hazard seismic zone requirements when properly mounted
- Recommended for indoor or outdoor bulk cylinder storage
- Surround cylinders with 2-in (51-mm) square steel structural tubing
- Continuously seam welded at all junction points
- Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
- Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
- For seismic installations, add Model 35350 S4-rated anchor bolts (not included)
Compliance
OSHA, CGA P1, NFPA55
Warranty
1 YEAR
Specifications
Steel | |
Black | |
Gas cylinder | |
762 mm |