missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Gas Cylinder Stand

Justrite™ Gas Cylinder Stand is designed for the safe storage of industrial and commercial use compressed gas cylinders. Help prevent dangerous tipping situations

Supplier:  Justrite™ JMC35280

 View more versions of this product

Catalog No. 19-085-826


Explore available promotions

Description

Description

  • Fully welded construction using steel or stainless steel provides structural integrity and longevity
  • Exclusive bumper guard edging with steel reinforced vinyl protects your cylinders from damage while providing extra grip
  • Accepts cylinders from 4 to 12in (102 to 305mm) in diameter
  • Pre-drilled holes in base for floor mounting with application specific fasteners (not included)
  • Heavy-duty polypropylene support straps, 1.5 x 54in (38mm x 1372mm) and rated at 1138lb (516kg), feature steel cinch buckle and hold individual cylinders securely against integral bracket
  • For an improved level of safety for any installation, all brackets include openings to retrofit secondary support chains, one set for each cylinder strap location

Compliance

OSHA, CGA P1, NFPA55

Warranty

1 YEAR

Specifications

Specifications

Gas Cylinder Stand
Stainless Steel
47 x 34 x 27 in.
762 mm
15 lb.
1 Cylinder
Silver
Gas cylinders
343 mm
483 mm
Videos
SDS
Documents

Documents

Promotions

Promotions