Thermo Scientific™ Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
$402.05 - $939.45
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
3162639
|
Thermo Scientific™
3162639 |
Small Gassing Hood | Precision 15L Shaking Water Baths |
Each for $543.27
|
|
||||
3162640
|
Thermo Scientific™
3162640 |
Large Gassing Hood | Precision 15L Shaking Water Baths |
Each for $764.89
|
|
||||
TSGPACL02
|
Thermo Scientific™
TSGPACL02 |
Clear Polycarbonate Gable Cover | Precision 2L General Purpose Water Bath |
Each for $402.05
|
|
||||
TSGPACL05
|
Thermo Scientific™
TSGPACL05 |
Clear Polycarbonate Gable Cover | Precision 2L and 5L General Purpose Water Baths |
Each for $466.79
|
|
||||
TSGPACL10
|
Thermo Scientific™
TSGPACL10 |
Clear Polycarbonate Gable Cover | Precision 10L General Purpose Water Baths |
Each for $527.57
|
|
||||
TSGPACL20
|
Thermo Scientific™
TSGPACL20 |
Clear Polycarbonate Gable Cover | Precision 20L and 28L General Purpose Water Baths |
Each for $590.34
|
|
||||
GPSSL02
|
Thermo Scientific™
GPSSL02 |
Stainless Steel Gable Cover | Precision 2L General Purpose Water Bath |
Each for $778.63
|
|
||||
GPSSL05
|
Thermo Scientific™
GPSSL05 |
Stainless Steel Gable Cover | Precision 2L and 5L General Purpose Water Baths |
Each for $849.21
|
|
||||
GPSSL10
|
Thermo Scientific™
GPSSL10 |
Stainless Steel Gable Cover | Precision 10L General Purpose Water Baths |
Each for $853.14
|
|
||||
GPSSL20
|
Thermo Scientific™
GPSSL20 |
Stainless Steel Gable Cover | Precision 20L and 28L General Purpose Water Baths |
Each for $939.45
|
|
||||
Description
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods- Permit control over the atmosphere surrounding the sample
- Improve temperature uniformity and reduce energy consumption
- Polished stainless steel with plastic gas fitting
- Each bath will accommodate 1 large hood or 2 small hoods
- Enhanced hinged design supports open lid configuration and easy lid removal
- Accommodate taller labware with extended height
- Polycarbonate and stainless steel models available
- Warranty varies by country, please contact us for details
Warranty
1 year