Learn More
Thermo Scientific™ Manual, Fixed Needle, Gas Tight Syringes for GC Instruments
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for manual accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
$158.77 - $642.66
Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Length (Metric) Needle | Needle Point Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Length (Metric) Needle | Needle Point Style | Price | Quantity | |||
365F2441
|
Thermo Scientific™
365F2441 |
25 μL | 23s G | 57 mm | Cone |
Each for $356.49
|
|
|||
365F2461
|
Thermo Scientific™
365F2461 |
25 μL | 26s G | 57 mm | Cone |
Each for $370.11
|
|
|||
365G2311
|
Thermo Scientific™
365G2311 |
50 μL | 23s G | 57 mm | Cone |
Each for $370.11
|
|
|||
365G2321
|
Thermo Scientific™
365G2321 |
50 μL | 26s G | 85 mm | Cone |
Each for $457.98
|
|
|||
365I2321
|
Thermo Scientific™
365I2321 |
250 μL | 26 G | 85 mm | Cone |
Each for $489.91
|
|
|||
36-5G2-111
|
Thermo Scientific™
365G2111 |
50 μL | 25 G | 50 mm | Bevel |
Each for $158.77
|
|
|||
365I2331
|
Thermo Scientific™
365I2331 |
250 μL | 26 G | 57 mm | Cone |
Each for $386.08
|
|
|||
365I2341
|
Thermo Scientific™
365I2341 |
250 μL | 23 G | 85 mm | Side Hole |
Each for $642.66
|
|
|||
36-5I2-561
|
Thermo Scientific™
365I2561 |
250 μL | 25 G | 50 mm | Bevel |
Each for $173.77
|
|
|||
36-5J2-831
|
Thermo Scientific™
365J2831 |
500 μL | 25 G | 50 mm | Bevel |
Each for $163.05
|
|
|||
36-5K3-051
|
Thermo Scientific™
365K3051 |
1 mL | 22 G | 50 mm | Bevel |
Each for $191.45
|
|
|||
03-100-053
|
Thermo Scientific™
365D6314 |
10 μL | 26s G | 50 mm | Blunt |
Each for $207.19
|
|
|||
03-100-056
|
Thermo Scientific™
365F6315 |
25 μL | 22s G | 50 mm | Blunt |
Each for $168.60
|
|
|||
03-100-057
|
Thermo Scientific™
365G6316 |
50 μL | 22s G | 50 mm | Blunt |
Each for $179.02
|
|
|||
03-100-058
|
Thermo Scientific™
365H6317 |
100 μL | 22s G | 50 mm | Blunt |
Each for $269.96
|
|
|||
03-100-059
|
Thermo Scientific™
365J6319 |
500 μL | 22s G | 50 mm | Blunt |
Each for $190.74
|
|
|||
Description
Get durability, clarity and accuracy from Thermo Scientific™ Manual Fixed Needle, Gas Tight Syringes. Used for manual sample injection into gas chromatography (GC) instrument, they provide accuracy and reproducibility that gives you confidence in analytical results, time-after-time. These syringes have an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes and types. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
- Borosilicate glass with metal flanges
- Needles are stainless steel
- Plungers are PTFE
- Field-proven syringe design
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.