Learn More
Therapak™ Gastroenterology (GI) Biopsy Collection and Transport Kits
Features standardized components to maximize the ease of collecting and packing specimens for transport
Supplier:  Therapak™ 38873
 View more versions of this product
Catalog No. 22-131-461
Only null left
Explore available promotions
Description
Description
Primary receptacles in the form of 20mL jars are 95kPa certified for transport in compliance with DOT and IATA standards for leak resistant transportation. A set of jar labels with the location ID is included in the kit to ensure proper placement of the specimen jars at the point of packing and identification once received at the test location. Customization of these kits is available upon request.
  • Comes with either two or six 20mL jars pre-filled with 10mL of formalin solution
  • Jars are secured by a corrugated insert and placed into the provided zip-style biohazard bag with absorbent material for transport
  • Corrugated inserts are printed with the GI biopsy location ID for easy recognition
Specifications
Specifications
2-Vial GI Biopsy Transport Kit
20/Cs.
Product Suggestions
Product Suggestions