Accuform Signs Gate Valve Lockouts
Available in five sizes these valve covers ensure no one will be able to operate valve handles
Supplier:  Accuform Signs KDD474
 View more versions of this product
Catalog No. 19-804-546
Explore available promotions
Description
Description
Specifications
Specifications
Gate Valve Lockout
Polypropylene
25 to 63 cm
−50° to +360°F
Red
10 to 13 in.
To enclose and fully surround the valve handle
−46° to +182°C