Cytiva Disposable PD-10 Columns
Use for gravity flow purification. Disposable PD-10 Columns are economical, plastic columns that can be packed with a wide range of bulk media.
Supplier:  Cytiva 17043501
Catalog No. 45-000-067
Explore available promotions
Description
Description
  • Economical and versatile columns for gravity flow applications
  • Empty PD-10 columns can be packed with a wide range of bulk media
  • Easy to use
  • Use PD-10 Columns with LabMate Buffer Reservoir for convenient equilibration, sample application, and wash
  • Empty Disposable PD-10 Columns are economical, plastic columns for single-use applications using gravity flow
  • The columns hold up to approx. 13 ml of chromatography media; the frits can be used as an upper bed support that prevents the medium from running dry under gravity flow
  • Comes with caps
Specifications
Specifications
Empty Columns, with Caps
Desalting
50 Pk.
0.59 in.
1.5 cm
Product Suggestions
Product Suggestions